Learn More
Aat Bioquest FMOC-Lys(5-FAM)-OH *CAS 1242933-88-5*
SDP
Supplier:  Aat Bioquest 5043
Encompass_Preferred
FMOC-Lys(5-FAM)-OH is a building block for in-sequence Lys labeling by FAM. FAM is one of the most common donor dyes for preparing FRET peptides (often paired with TAMRA or TQ2).
Catalog No. 50-195-3260