Learn More
Bachem Fmoc-Lys(Aloc)-OH
SDP
Supplier:  Bachem 4016656.0005
Encompass_Preferred
Fmoc-Lys(Aloc)-OH, 146982-27-6, C25H28N2O6, Mr 452.51, 5g. The selectively cleavable Alloc group allows on-resin modification of lysine side-chains during Fmoc-SPPS.
Catalog No. 50-312-763