Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Fmoc-Lys(Boc)-Oh

Catalog No. 4762410GF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-624-10GF 10 g
47-624-100GF 100 g
47-624-25GF 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 47-624-10GF Supplier Sigma Aldrich Supplier No. 4762410GF
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Fmoc-Lys(Boc)-OH can be used for:The solid-phase synthesis of pentasubstituted dihydroimidazolylbutyl dihydroimidazol-3-ium salts by coupling with p-methylbenzhydrylamine (MBHA) resin.Fmoc-based peptide synthesis of bis-naphthalene diimide, a threading intercalator.Synthesis of +a-Boc-+a-(3,5-bis-trifluoromethyl-benzyl)-+a-Fmoc-L-Lysine to be used as 19F NMR based screening tool.

Specifications

CAS 71989-26-9
Molecular Formula C26H32N2O6
Linear Formula C26H32N2O6
MDL Number MFCD00037138
Quantity 10 g
Synonym Nalpha-Fmoc-Nepsilon-Boc-L-lysine
Molecular Weight (g/mol) 468.54 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.