Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fmoc-Lys Boc -OH, Advanced ChemTech

Catalog No. 01340276
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-340-276 100 g
01-340-272 5 g
01-340-273 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-340-276 Supplier Advanced Chem Tech Supplier No. FK2390100G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 71989-26-9 | C26H32N2O6 | 468.55 g/mol

Chemical Identifiers

CAS 71989-26-9
Molecular Formula C26H32N2O6
Molecular Weight (g/mol) 468.55
MDL Number MFCD000371
InChI Key UMRUUWFGLGNQLI-ANBDAQEENA-N
IUPAC Name (2S)-6-{[(tert-butoxy)carbonyl]amino}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid
SMILES CC(C)(C)OC(=O)NCCCC[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O

Specifications

Color White/Off-White
Quantity 100 g
Grade Biochemical
Chemical Name or Material Fmoc-Lys Boc -OH
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.