Learn More
MilliporeSigma FMOC-LYS(BOC)-SER(PSIME MEPRO)
Catalog No. 502303089
Encompass
Fmoc-Lys(Boc)-Ser(PsiMeMepro)-OH; > /=97% by HPLC; White to light yellow solid crystalline powder; EMD; Novabiochem; F.W. 595.7; C32H41N3O8
Catalog No. 50-230-3089 Supplier MilliporeSigma Supplier No. 8521780005
Add to Cart
Add to Cart
Fmoc-Lys(Boc)-Ser(PsiMeMepro)-OH; > /=97% by HPLC; White to light yellow solid crystalline powder; EMD; Novabiochem; F.W. 595.7; C32H41N3O8