Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fmoc-Lys Boc -Thr psiMe.Mepro , Advanced ChemTech

Catalog No. 01339388
Click to view available options
Quantity:
1 g
5 g

Catalog No. 01-339-388

Supplier: Advanced Chem Tech PTK0231G

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 911838-56-7
Molecular Formula C33H43N3O8
Molecular Weight (g/mol) 609.72
MDL Number MFCD11974987
InChI Key JAHDJVRHKPYPNY-PHXCCWLDSA-N
IUPAC Name (4S,5R)-3-[(2S)-6-{[(tert-butoxy)carbonyl]amino}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoyl]-2,2,5-trimethyl-1,3-oxazolidine-4-carboxylic acid
SMILES C[C@H]1OC(C)(C)N([C@@H]1C(O)=O)C(=O)[C@H](CCCCNC(=O)OC(C)(C)C)NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2

Specifications

Color White/Off-White
Quantity 1 g
Grade Biochemical
Chemical Name or Material Fmoc-Lys Boc -Thr psiMe.Mepro
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.