Learn More
Bachem Fmoc-Lys(Boc)-Wang resin (100-200 mesh, 0.5-0.8 mmol/g)
SDP
Catalog No. 50258922
Encompass_Preferred
Fmoc-Lys(Boc)-Wang resin (100-200 mesh, 0.5-0.8 mmol/g), Fmoc-Lys(Boc)-4-alkoxybenzyl alcohol resin, 5g. Fmoc-Lys(Boc)-p-alkoxybenzyl alcohol resin.
Catalog No. 50-258-922 Supplier Bachem Supplier No. 4028589.0005
Fmoc-Lys(Boc)-Wang resin (100-200 mesh, 0.5-0.8 mmol/g), Fmoc-Lys(Boc)-4-alkoxybenzyl alcohol resin, 5g. Fmoc-Lys(Boc)-p-alkoxybenzyl alcohol resin.