Learn More
AnaSpec FMOC-LYS(DABCYL)-OH
Supplier:  AnaSpec AS2349601
Encompass
Fmoc-Lys(DABCYL)-OH[N-a -Fmoc-N-E -(4,4-dimethylazobenzene-4’carbonyl)-L-lysine] ; MW 619.71; M.F. C36H37N5O5 (100 mg) A useful building block for FRET probes containing DABCYL (as acceptor).
Catalog No. 50350588