Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fmoc-Lys Fmoc 4-Lys2-Lys-Cys, Advanced ChemTech
Click to view available options
:
Quantity: 1000 mg
Specifications
Specifications
| Color | White/Off-White |
| Quantity | 1000 mg |
| Grade | Biochemical |
| Chemical Name or Material | Fmoc-Lys Fmoc 4-Lys2-Lys-Cys |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction