Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fmoc-Lys(Fmoc)-OH | 78081-87-5 | 590.66 | 500 G
SDP

Catalog No. 5000447302
Encompass_Preferred

Fmoc-Lys(Fmoc)-OH is a lysine derivative used commercially as an ergogenic supplement. Amino acids and their derivatives influence anabolic hormone secretion, fuel supply during exercise, mental performance under stress, and can prevent exercise-induced muscle damage. This product is recognized as a beneficial ergogenic dietary substance and is for research use only.

  • Lysine derivative
  • Influences anabolic hormone secretion
  • Supports fuel supply during exercise
  • Enhances mental performance under stress
  • Helps prevent exercise-induced muscle damage
  • High purity of 99.22%
  • Molecular weight of 590.66
  • Formula: C36H34N2O6
  • Appearance: Solid, white to off-white
  • Recommended storage at room temperature for up to 3 years; in solvent, at -80°C for 2 years or -20°C for 1 year

Catalog No. 50-004-47302 Supplier Medchemexpress LLC Supplier No. HYW008386500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-Lys(Fmoc)-OH is a lysine derivative used commercially as an ergogenic supplement. Amino acids and their derivatives influence anabolic hormone secretion, fuel supply during exercise, mental performance under stress, and can prevent exercise-induced muscle damage. This product is recognized as a beneficial ergogenic dietary substance and is for research use only.

  • Lysine derivative
  • Influences anabolic hormone secretion
  • Supports fuel supply during exercise
  • Enhances mental performance under stress
  • Helps prevent exercise-induced muscle damage
  • High purity of 99.22%
  • Molecular weight of 590.66
  • Formula: C36H34N2O6
  • Appearance: Solid, white to off-white
  • Recommended storage at room temperature for up to 3 years; in solvent, at -80°C for 2 years or -20°C for 1 year

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.