Learn More
AnaSpec FMOC-LYS(IVDDE)-OH
Supplier:  AnaSpec AS629345
Encompass
Fmoc-Lys(ivDde)-OH, ; MW 574.6; M.F. C34H42N2O6 (5 g) HPLC: purity:96%. Optical purity: <=0.50% D-enantiomer. Solubility: 1 mmole in 2 ml DMF clearly solube.
Catalog No. 50352435