Learn More
AnaSpec FMOC-LYS(N3)-OH 250MG
Catalog No. 50352210
Encompass
Fmoc-Lys(N3)-OH[Fmoc-azidolysine; Fmoc-lys(azide); Fmoc-lysine azide] ; MW 394.42; M.F. C21H22N4O4 (250 mg) An unusual amino acid used in Solid Phase Peptide Synthesis (SPPS) for the synthesis of side-chain modified peptides and proteins.1 The side-chain azido (N3) group is stable in trifluoroacetic acid (TFA) or piperidine (Pip), two reagents commonly used in SPPS. The azido group can be readily converted to an amine on the solid phase or in solution via the Staudinger reduction.2
Catalog No. 50352210 Supplier AnaSpec Supplier No. AS53100F025
Fmoc-Lys(N3)-OH[Fmoc-azidolysine; Fmoc-lys(azide); Fmoc-lysine azide] ; MW 394.42; M.F. C21H22N4O4 (250 mg) An unusual amino acid used in Solid Phase Peptide Synthesis (SPPS) for the synthesis of side-chain modified peptides and proteins.1 The side-chain azido (N3) group is stable in trifluoroacetic acid (TFA) or piperidine (Pip), two reagents commonly used in SPPS. The azido group can be readily converted to an amine on the solid phase or in solution via the Staudinger reduction.2