Learn More
Bachem Fmoc-Lys-OH · HCl
SDP
Supplier:  Bachem 4014974.0001
Encompass_Preferred
Fmoc-Lys-OH · HCl, 139262-23-0, C21H24N2O4 · HCl, Mr 404.89, 1g. Fmoc-Lys-OH has been converted into the e-azido derivative using triflic azide in the presence of CuSO4 and NaHCO3.
Catalog No. 50-312-704