Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fmoc-Lys(palmitoyl)-OH | 201004-46-8 | MFCD00237026 | >97.0% | 606.84 g/mol | C37H54N2O5 | 1 G
SDP

Catalog No. 5000209757
Encompass_Preferred

Fmoc-Lys(palmitoyl)-OH is an Fmoc-protected amino acid in which the lysine side chain is modified with a palmitoyl (C16) acyl group. It is supplied as a white to off-white solid and is intended for use in solid-phase peptide synthesis to introduce lipidation (palmitoylation) on lysine residues for research applications.

  • White to off-white solid with high purity.
  • Purity greater than 97.0% as determined by NMR.
  • Molecular weight 606.84 g/mol and formula C37H54N2O5.
  • Suitable for solid-phase peptide synthesis and lipidated peptide design.
  • Store powder at -20°C for long-term stability; short-term storage at 4°C is acceptable.

Catalog No. 50-002-09757 Supplier Medchemexpress LLC Supplier No. HYW0482091G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-Lys(palmitoyl)-OH is an Fmoc-protected amino acid in which the lysine side chain is modified with a palmitoyl (C16) acyl group. It is supplied as a white to off-white solid and is intended for use in solid-phase peptide synthesis to introduce lipidation (palmitoylation) on lysine residues for research applications.

  • White to off-white solid with high purity.
  • Purity greater than 97.0% as determined by NMR.
  • Molecular weight 606.84 g/mol and formula C37H54N2O5.
  • Suitable for solid-phase peptide synthesis and lipidated peptide design.
  • Store powder at -20°C for long-term stability; short-term storage at 4°C is acceptable.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.