Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fmoc-lys(palmitoyl)-OH | 201004-46-8 | MFCD00237026 | >97.0% | 606.84 g/mol | C37H54N2O5 | 5 G
SDP

Catalog No. 5000210304
Encompass_Preferred

Fmoc-lys(palmitoyl)-OH is an Fmoc-protected lysine derivative where the lysine side-chain amine is acylated with a palmitoyl (C16) lipid chain. It is supplied as a white to off-white solid amino-acid building block intended for incorporation of lipidated residues during solid-phase peptide synthesis. The material is for research use only and is supplied with certificate of analysis information regarding purity and storage.

  • Fmoc-protected lysine with palmitoyl side-chain.
  • Used as a building block for solid-phase peptide synthesis.
  • Molecular weight about 606.84 g/mol, formula C37H54N2O5.
  • Purity ≥97.0% by NMR.
  • Appearance: white to off-white solid.
  • Recommended storage: powder at -20°C (3 years) or 4°C (2 years); in solvent at -80°C (6 months) or -20°C (1 month).
  • For research use only; not for human therapeutic use.

Catalog No. 50-002-10304 Supplier Medchemexpress LLC Supplier No. HYW0482095G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-lys(palmitoyl)-OH is an Fmoc-protected lysine derivative where the lysine side-chain amine is acylated with a palmitoyl (C16) lipid chain. It is supplied as a white to off-white solid amino-acid building block intended for incorporation of lipidated residues during solid-phase peptide synthesis. The material is for research use only and is supplied with certificate of analysis information regarding purity and storage.

  • Fmoc-protected lysine with palmitoyl side-chain.
  • Used as a building block for solid-phase peptide synthesis.
  • Molecular weight about 606.84 g/mol, formula C37H54N2O5.
  • Purity ≥97.0% by NMR.
  • Appearance: white to off-white solid.
  • Recommended storage: powder at -20°C (3 years) or 4°C (2 years); in solvent at -80°C (6 months) or -20°C (1 month).
  • For research use only; not for human therapeutic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.