Learn More
Medchemexpress LLC Fmoc-Met-OH-13C5,15N | 1217437-64-3 | 377.41 | 5 MG
SDP
Catalog No. 5000447313
Encompass_Preferred
Fmoc-Met-OH-13C5,15N is a 15N-labeled and 13C-labeled Fmoc-Met-OH. This product is a Methionine derivative. Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into drug molecules, primarily as tracers for quantitation during drug development. Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
  • 15N-labeled and 13C-labeled compound
  • Methionine derivative
  • Used as tracers for quantitation during drug development
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-47313 Supplier Medchemexpress LLC Supplier No. HYW008371S15MG
Fmoc-Met-OH-13C5,15N is a 15N-labeled and 13C-labeled Fmoc-Met-OH. This product is a Methionine derivative. Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into drug molecules, primarily as tracers for quantitation during drug development. Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
  • 15N-labeled and 13C-labeled compound
  • Methionine derivative
  • Used as tracers for quantitation during drug development
  • Potential to affect pharmacokinetic and metabolic profiles of drugs