Learn More
Medchemexpress LLC Fmoc-Met-OH-15N | 934183-50-3 | MFCD04118239 | 372.44 | 10 MG
SDP
Catalog No. 5000447305
Encompass_Preferred
Fmoc-Met-OH-15N is a methionine derivative, specifically an N-protected amino acid with a 15N isotope, ideal for peptide synthesis. It features a fluorenylmethoxycarbonyl (Fmoc) group for protection and is suitable for various analytical techniques, including bio NMR. This product appears as a white to off-white solid.
  • Suitable for bio NMR technique
  • Applied in peptide synthesis
  • Features an Fmoc functional group
  • Exhibits a mass shift of M+1
  • High purity of 99% (HPLC)
  • Isotopic enrichment of 99%
Catalog No. 50-004-47305 Supplier Medchemexpress LLC Supplier No. HYW008371S10MG
Fmoc-Met-OH-15N is a methionine derivative, specifically an N-protected amino acid with a 15N isotope, ideal for peptide synthesis. It features a fluorenylmethoxycarbonyl (Fmoc) group for protection and is suitable for various analytical techniques, including bio NMR. This product appears as a white to off-white solid.
  • Suitable for bio NMR technique
  • Applied in peptide synthesis
  • Features an Fmoc functional group
  • Exhibits a mass shift of M+1
  • High purity of 99% (HPLC)
  • Isotopic enrichment of 99%