Learn More
Apexbio Technology LLC Fmoc-Met-OH 71989-28-1 500g
SDP
Catalog No. 5000092553
Encompass_Preferred
Fmoc-Met-OH (71989-28-1) is a derivative of the amino acid methionine protected by the Fmoc (9-fluorenylmethoxycarbonyl) group and is extensively used in solid-phase peptide synthesis (SPPS) Fmoc-Met-OH is specifically utilized in the synthesis of methionine-containing peptides for biomedical research including the production of bioactive peptides pharmaceutical precursor compounds and investigation of protein structure-function relationships Its compatibility with solid-phase techniques enables efficient assembly of complex peptide sequences for further biological analysis and therapeutic studies
Catalog No. 50-000-92553 Supplier Apexbio Technology LLC Supplier No. A7939500000
Fmoc-Met-OH (71989-28-1) is a derivative of the amino acid methionine protected by the Fmoc (9-fluorenylmethoxycarbonyl) group and is extensively used in solid-phase peptide synthesis (SPPS) Fmoc-Met-OH is specifically utilized in the synthesis of methionine-containing peptides for biomedical research including the production of bioactive peptides pharmaceutical precursor compounds and investigation of protein structure-function relationships Its compatibility with solid-phase techniques enables efficient assembly of complex peptide sequences for further biological analysis and therapeutic studies