Learn More
Medchemexpress LLC Fmoc-Met-OH-d3 | 502692-58-2 | 374.47 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW008371S210MG
Encompass_Preferred
Fmoc-Met-OH-d3 is the deuterium labeled Fmoc-Met-OH. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled compound.
  • Functions as a tracer.
  • Acts as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes into drug molecules.
  • Potentially affects pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-47437