Learn More
Medchemexpress LLC Fmoc-Met-OH-d3 | 502692-58-2 | 374.47 | 5 MG
SDP
Catalog No. 5000447304
Encompass_Preferred
Fmoc-Met-OH-d3 is the deuterium labeled Fmoc-Met-OH. It serves as a tracer and can be used as an internal standard for quantitative analysis.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR
  • Can be used as an internal standard for quantitative analysis by GC-MS
  • Can be used as an internal standard for quantitative analysis by LC-MS
Catalog No. 50-004-47304 Supplier Medchemexpress LLC Supplier No. HYW008371S25MG
Fmoc-Met-OH-d3 is the deuterium labeled Fmoc-Met-OH. It serves as a tracer and can be used as an internal standard for quantitative analysis.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR
  • Can be used as an internal standard for quantitative analysis by GC-MS
  • Can be used as an internal standard for quantitative analysis by LC-MS