Learn More
Broadpharm Fmoc-N-Amido-PEG8-Acid, 5 g, CAS 756526-02-0, Solid or viscous liquid, 98% purity, Linear, dPEG spacer, 8 PEG units
Catalog No. NC3302003
Encompass
Fmoc-N-Amido-PEG8-Acid is a versatile heterobifunctional polyethylene glycol (PEG) linker specifically designed for applications in bioconjugation, including peptide synthesis and Antibody-Drug Conjugates (ADCs). This product features a unique Fmoc-protected amine and a terminal carboxylic acid, offering two anchor points for modifications. The hydrophilic PEG8 spacer enhances solubility in aqueous media, improving the pharmacokinetic properties of peptides and extending their circulation times in vivo.
  • Designed for versatile modifications with two distinct functional groups.
  • Improves water solubility and expands hydrodynamic volume.
  • Reduces renal clearance and protects peptides from proteolysis.
  • Facilitates stable amide bond formation with primary amines using activators.
  • Stable under -20°C storage conditions.
  • Shipping is done at ambient temperature.
Catalog No. NC3302003 Supplier Broadpharm Supplier No. BP216315G
Fmoc-N-Amido-PEG8-Acid is a versatile heterobifunctional polyethylene glycol (PEG) linker specifically designed for applications in bioconjugation, including peptide synthesis and Antibody-Drug Conjugates (ADCs). This product features a unique Fmoc-protected amine and a terminal carboxylic acid, offering two anchor points for modifications. The hydrophilic PEG8 spacer enhances solubility in aqueous media, improving the pharmacokinetic properties of peptides and extending their circulation times in vivo.
  • Designed for versatile modifications with two distinct functional groups.
  • Improves water solubility and expands hydrodynamic volume.
  • Reduces renal clearance and protects peptides from proteolysis.
  • Facilitates stable amide bond formation with primary amines using activators.
  • Stable under -20°C storage conditions.
  • Shipping is done at ambient temperature.