Learn More
Medchemexpress LLC Fmoc-N-Me-Ala-OH | 84000-07-7 | 325.36 | 50 G
SDP
Catalog No. 5000448750
Encompass_Preferred
Fmoc-N-Me-Ala-OH is an Fmoc-protected N-methyl-L-alanine that serves as a building block for solid-phase peptide synthesis. It is utilized to synthesize peptidomimetic antagonists of XIAP BIR3 derived from Smac/DIABLO.
  • Can be incorporated into peptidomimetic compounds that restore caspase-9 activity.
  • Peptidomimetic compounds incorporating this chemical show moderate cytotoxicity against MDA-MB-231 and MCF-7 breast cancer cell lines.
Catalog No. 50-004-48750 Supplier Medchemexpress LLC Supplier No. HYW00823550G
Fmoc-N-Me-Ala-OH is an Fmoc-protected N-methyl-L-alanine that serves as a building block for solid-phase peptide synthesis. It is utilized to synthesize peptidomimetic antagonists of XIAP BIR3 derived from Smac/DIABLO.
  • Can be incorporated into peptidomimetic compounds that restore caspase-9 activity.
  • Peptidomimetic compounds incorporating this chemical show moderate cytotoxicity against MDA-MB-231 and MCF-7 breast cancer cell lines.