Learn More
Medchemexpress LLC Fmoc-N-Me-Arg(Pbf)-OH | 913733-27-4 | 98.3% | 662.80 | 500 MG
SDP
Catalog No. 5000350714
Encompass_Preferred
Fmoc-N-Me-Arg(Pbf)-OH is an amino acid derivative that contains a guanidinium protecting group on the arginine side chain. It is utilized in the synthesis of neurotensin-derived NTS1 ligands for PET imaging.
  • Amino acid derivative
  • Contains a guanidinium protecting group
  • Used in synthesis of neurotensin-derived NTS1 ligands
  • Suitable for PET imaging
  • Appears as a solid
  • White to off-white color
Catalog No. 50-003-50714 Supplier Medchemexpress LLC Supplier No. HY137002500MG
Fmoc-N-Me-Arg(Pbf)-OH is an amino acid derivative that contains a guanidinium protecting group on the arginine side chain. It is utilized in the synthesis of neurotensin-derived NTS1 ligands for PET imaging.
  • Amino acid derivative
  • Contains a guanidinium protecting group
  • Used in synthesis of neurotensin-derived NTS1 ligands
  • Suitable for PET imaging
  • Appears as a solid
  • White to off-white color