Learn More
Medchemexpress LLC Fmoc-N-Me-D-Arg Pbf | MFCD04974250 | 99.6% | 662.80 | C35H42N4O7S | 50 MG
SDP
Catalog No. 5000282863
Encompass_Preferred
Fmoc-N-Me-D-Arg(Pbf)-OH is an Fmoc-protected, N-methylated D-arginine derivative supplied as a protected amino acid building block for solid-phase peptide synthesis and related research. The material features Pbf side-chain protection for the guanidino group, is provided in high purity, and is supported by manufacturer analytical and safety documents.
  • Fmoc-protected N-methyl D-arginine for peptide synthesis.
  • Pbf side-chain protection compatible with common deprotection protocols.
  • High purity (99.6%) for reliable synthetic performance.
  • Available as solid and as a 10 mM solution in DMSO.
  • Multiple package sizes to support small-scale and larger projects.
  • Manufacturer-provided analytical data: COA, HNMR, LC-MS, RP-HPLC.
  • Safety data and handling instructions available.
Catalog No. 50-002-82863 Supplier Medchemexpress LLC Supplier No. HYW14576250MG
Fmoc-N-Me-D-Arg(Pbf)-OH is an Fmoc-protected, N-methylated D-arginine derivative supplied as a protected amino acid building block for solid-phase peptide synthesis and related research. The material features Pbf side-chain protection for the guanidino group, is provided in high purity, and is supported by manufacturer analytical and safety documents.
  • Fmoc-protected N-methyl D-arginine for peptide synthesis.
  • Pbf side-chain protection compatible with common deprotection protocols.
  • High purity (99.6%) for reliable synthetic performance.
  • Available as solid and as a 10 mM solution in DMSO.
  • Multiple package sizes to support small-scale and larger projects.
  • Manufacturer-provided analytical data: COA, HNMR, LC-MS, RP-HPLC.
  • Safety data and handling instructions available.