Learn More
Medchemexpress LLC Fmoc-N-Me-D-Arg(Pbf)-OH | 2044710-57-6 | MFCD04974250 | 99.6% | 662.80 g·mol⁻1 | C35H42N4O7S | 25 MG
SDP
Catalog No. 5000282862
Encompass_Preferred
Fmoc-N-Me-D-Arg(Pbf)-OH is an Fmoc-protected, N-methylated D-arginine derivative bearing a Pbf side-chain protecting group. Supplied as a white to off-white solid with high reported purity, it is used as a protected amino acid building block in solid-phase peptide synthesis to introduce N-methyl D-arginine residues for SAR studies and peptide modification.
  • Fmoc-protected N-methyl D-arginine suitable for solid-phase peptide synthesis.
  • Pbf side-chain protection compatible with standard acidic deprotection.
  • High purity for consistent synthetic performance.
  • Available in small-scale vial and solution formats for flexibility.
  • Store powder at -20°C for long-term stability; follow solvent storage recommendations.
Catalog No. 50-002-82862 Supplier Medchemexpress LLC Supplier No. HYW14576225MG
Fmoc-N-Me-D-Arg(Pbf)-OH is an Fmoc-protected, N-methylated D-arginine derivative bearing a Pbf side-chain protecting group. Supplied as a white to off-white solid with high reported purity, it is used as a protected amino acid building block in solid-phase peptide synthesis to introduce N-methyl D-arginine residues for SAR studies and peptide modification.
  • Fmoc-protected N-methyl D-arginine suitable for solid-phase peptide synthesis.
  • Pbf side-chain protection compatible with standard acidic deprotection.
  • High purity for consistent synthetic performance.
  • Available in small-scale vial and solution formats for flexibility.
  • Store powder at -20°C for long-term stability; follow solvent storage recommendations.