Learn More
Medchemexpress LLC Fmoc-N-Me-D-Trp Boc 5g
SDP
Catalog No. 5000204915
Encompass_Preferred
Fmoc-N-Me-D-Trp(Boc)-OH is an amino acid derivative with an Fmoc protecting group which can be used to synthesize somatostatin-dopamine chimeric analogs[1]
Catalog No. 50-002-04915 Supplier Medchemexpress LLC Supplier No. HYW1063255G
Fmoc-N-Me-D-Trp(Boc)-OH is an amino acid derivative with an Fmoc protecting group which can be used to synthesize somatostatin-dopamine chimeric analogs[1]