Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fmoc-N-Me-Glu(OtBu)-OH | 200616-40-6 | 99.7% | 439.50 | 5 G
SDP

Catalog No. 5000395585
Encompass_Preferred

Fmoc-N-Me-Glu(OtBu)-OH is a glutamic acid derivative. Amino acids and their derivatives are widely used as ergogenic supplements, influencing the secretion of anabolic hormones, supplying fuel during exercise, enhancing mental performance during stress, and preventing exercise-induced muscle damage.

  • Purity of 99.67%
  • Appears as a white to off-white solid
  • Molecular weight: 439.50
  • For research use only

Catalog No. 50-003-95585 Supplier Medchemexpress LLC Supplier No. HYW0085295G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-N-Me-Glu(OtBu)-OH is a glutamic acid derivative. Amino acids and their derivatives are widely used as ergogenic supplements, influencing the secretion of anabolic hormones, supplying fuel during exercise, enhancing mental performance during stress, and preventing exercise-induced muscle damage.

  • Purity of 99.67%
  • Appears as a white to off-white solid
  • Molecular weight: 439.50
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.