Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fmoc-N-Me-Ile-OH | 138775-22-1 | MFCD00153389 | 99.88% | 367.44 | 50 G
SDP

Catalog No. 5000423356
Encompass_Preferred

Fmoc-N-Me-Ile-OH is an amino acid derivative used in peptide synthesis.

  • Purity: 99.88%
  • Molecular weight: 367.44
  • Storage (powder): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month

Catalog No. 50-004-23356 Supplier Medchemexpress LLC Supplier No. HYW00855550G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-N-Me-Ile-OH is an amino acid derivative used in peptide synthesis.

  • Purity: 99.88%
  • Molecular weight: 367.44
  • Storage (powder): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.