Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fmoc-N-PEG20-acid | 1952360-93-8 | C58H97NO24 | 100 MG
SDP

Catalog No. 5000353067
Encompass_Preferred

Fmoc-N-amido-PEG20-acid is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs themselves are designed with two distinct ligands connected by a linker: one ligand targets an E3 ubiquitin ligase, and the other targets the protein of interest. This mechanism exploits the intracellular ubiquitin-proteasome system to facilitate the selective degradation of specific target proteins.

  • Peg-based protac linker
  • Used in the synthesis of protacs
  • Molecular weight: 1192.38
  • Appearance: Solid
  • Color: Light yellow to brown

Catalog No. 50-003-53067 Supplier Medchemexpress LLC Supplier No. HY140462100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Fmoc-N-amido-PEG20-acid is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs themselves are designed with two distinct ligands connected by a linker: one ligand targets an E3 ubiquitin ligase, and the other targets the protein of interest. This mechanism exploits the intracellular ubiquitin-proteasome system to facilitate the selective degradation of specific target proteins.

  • Peg-based protac linker
  • Used in the synthesis of protacs
  • Molecular weight: 1192.38
  • Appearance: Solid
  • Color: Light yellow to brown

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.