Learn More
Medchemexpress LLC Fmoc-N-PEG36-acid | 2649489-46-1 | C90H161NO40 | 25 MG
SDP
Catalog No. 5000355938
Encompass_Preferred
Fmoc-N-PEG36-acid is a polyethylene glycol (PEG)-based linker designed for laboratory applications, particularly in PROTAC synthesis. It is used for constructing bifunctional molecules that recruit target proteins to an E3 ubiquitin ligase for degradation, and is intended for research use only.
  • Keep container tightly sealed in a cool, well-ventilated area.
  • Protect from direct sunlight and sources of ignition.
  • Store powder at -20°C for up to 3 years.
  • Store in solvent at -80°C for 6 months or -20°C for 1 month.
  • Shipping at room temperature is acceptable for durations less than 2 weeks.
  • Harmful if swallowed.
  • May cause skin irritation.
  • May cause serious eye irritation.
  • May cause respiratory irritation.
Catalog No. 50-003-55938 Supplier Medchemexpress LLC Supplier No. 25MGHY140464
Add to Cart
Add to Cart
Fmoc-N-PEG36-acid is a polyethylene glycol (PEG)-based linker designed for laboratory applications, particularly in PROTAC synthesis. It is used for constructing bifunctional molecules that recruit target proteins to an E3 ubiquitin ligase for degradation, and is intended for research use only.
  • Keep container tightly sealed in a cool, well-ventilated area.
  • Protect from direct sunlight and sources of ignition.
  • Store powder at -20°C for up to 3 years.
  • Store in solvent at -80°C for 6 months or -20°C for 1 month.
  • Shipping at room temperature is acceptable for durations less than 2 weeks.
  • Harmful if swallowed.
  • May cause skin irritation.
  • May cause serious eye irritation.
  • May cause respiratory irritation.