Learn More
AnaSpec FMOC-NH-(PEG)2-CH2CH2COOH
Supplier:  AnaSpec AS650391
Encompass
Fmoc-NH-(PEG)2-CH2CH2COOH[Fmoc-9-Amino-4; 7-Dioxanonanoic acid; Fmoc-3-(2-(2-aminoethoxy)ethoxy)propanoic acid] ; MW 399.44; M.F. C22H25NO6 (1 g)
Catalog No. 50352542