Learn More
AnaSpec FMOC-NH-(PEG)5-CH2CH2COOH
Supplier:  AnaSpec AS650361
Encompass
Fmoc-NH-(PEG)5-CH2CH2COOH[Fmoc-18-amino-4,7,10,13,16-pentaoxaoctadecanoic acid] ; MW 531.59; M.F. C28H37NO9 (1 g)
Catalog No. 50352536