Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fmoc-NH-(PEG)6-CH2CH2COOH | 882847-34-9 | 99.3% | 575.65 | C30H41NO10 | 500 MG
SDP

Catalog No. 5000202326
Encompass_Preferred

Fmoc-NH-(PEG)6-CH2CH2COOH is an Fmoc-protected PEG6 propionic acid linker used as a cleavable linker in the synthesis of antibody-drug conjugates (ADCs). It is characterized by high purity, defined molecular properties, and recommended cold storage for stability.

  • Cleavable linker for antibody-drug conjugation.
  • Fmoc-protected amine functionality for stepwise synthesis.
  • PEG6 spacer provides hydrophilicity and flexibility.
  • High purity: 99.3%.
  • Molecular weight 575.65; formula C30H41NO10.
  • Available in multiple sizes: 100 mg, 500 mg, 1 g, 5 g, 10 g.
  • Appearance: colorless to light yellow liquid; density 1.202 ± 0.06 g/cm3.
  • Storage: -20°C (3 years) or 4°C (2 years) in pure form; in solution -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-02326 Supplier Medchemexpress LLC Supplier No. HYW040246500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-NH-(PEG)6-CH2CH2COOH is an Fmoc-protected PEG6 propionic acid linker used as a cleavable linker in the synthesis of antibody-drug conjugates (ADCs). It is characterized by high purity, defined molecular properties, and recommended cold storage for stability.

  • Cleavable linker for antibody-drug conjugation.
  • Fmoc-protected amine functionality for stepwise synthesis.
  • PEG6 spacer provides hydrophilicity and flexibility.
  • High purity: 99.3%.
  • Molecular weight 575.65; formula C30H41NO10.
  • Available in multiple sizes: 100 mg, 500 mg, 1 g, 5 g, 10 g.
  • Appearance: colorless to light yellow liquid; density 1.202 ± 0.06 g/cm3.
  • Storage: -20°C (3 years) or 4°C (2 years) in pure form; in solution -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.