Learn More
AnaSpec FMOC-NH-(PEG)8-CH2CH2COOH
Supplier:  AnaSpec AS650341
Encompass
Fmoc-NH-(PEG)8-CH2CH2COOH[1-(9H-fluoren-9-ylmethyl)ester; 5,8,11,14,17,20,23,26-Octaoxa-2-azanonacosanedioic acid] ; MW 663.76; M.F. C34H49NO12 (1 g)
Catalog No. 50352532