Learn More
Medchemexpress LLC Fmoc-NH-PEG1-CH2COOH | 260367-12-2 | 99.5% | 341.36 g/mol | C19H19NO5 | 1 G
SDP
Catalog No. 5000203035
Encompass_Preferred
Fmoc-NH-PEG1-CH2COOH is an Fmoc-protected PEG-based linker with a terminal carboxylic acid, designed for installation of linkers in medicinal chemistry workflows such as ADC and PROTAC synthesis.
  • Used in antibody-drug conjugate linker synthesis.
  • Used in proteolysis-targeting chimera (PROTAC) synthesis.
  • Contains an Fmoc-protected amine and a terminal carboxylic acid for conjugation.
  • White to off-white solid with good purity for research applications.
  • Available in multiple sizes, including 1 g, for flexible laboratory use.
Catalog No. 50-002-03035 Supplier Medchemexpress LLC Supplier No. HYW0558611G
Fmoc-NH-PEG1-CH2COOH is an Fmoc-protected PEG-based linker with a terminal carboxylic acid, designed for installation of linkers in medicinal chemistry workflows such as ADC and PROTAC synthesis.
  • Used in antibody-drug conjugate linker synthesis.
  • Used in proteolysis-targeting chimera (PROTAC) synthesis.
  • Contains an Fmoc-protected amine and a terminal carboxylic acid for conjugation.
  • White to off-white solid with good purity for research applications.
  • Available in multiple sizes, including 1 g, for flexible laboratory use.