Learn More
Medchemexpress LLC Fmoc-nh-peg1-ch2cooh | 260367-12-2 | MFCD06796876 | 99.9% | 341.36 | C19H19NO5 | 5 G
SDP
Catalog No. 5000202927
Encompass_Preferred
Fmoc-NH-PEG1-CH2COOH is an Fmoc-protected PEG1 linker used as a cleavable linker for antibody-drug conjugate (ADC) synthesis and as a PEG-based PROTAC linker. The compound is supplied as a white powder with reported high purity and is available in multiple pack sizes, including a 5 G pack.
  • Fmoc-protected amine with a PEG1 spacer and terminal carboxylic acid.
  • Used as a cleavable linker in ADC and PROTAC syntheses.
  • Supplied as a white powder; multiple pack sizes available.
  • Reported high purity suitable for research applications.
  • Chemical identifiers: CAS 260367-12-2; molecular weight 341.36; formula C19H19NO5.
Catalog No. 50-002-02927 Supplier Medchemexpress LLC Supplier No. HYW0558615G
Fmoc-NH-PEG1-CH2COOH is an Fmoc-protected PEG1 linker used as a cleavable linker for antibody-drug conjugate (ADC) synthesis and as a PEG-based PROTAC linker. The compound is supplied as a white powder with reported high purity and is available in multiple pack sizes, including a 5 G pack.
  • Fmoc-protected amine with a PEG1 spacer and terminal carboxylic acid.
  • Used as a cleavable linker in ADC and PROTAC syntheses.
  • Supplied as a white powder; multiple pack sizes available.
  • Reported high purity suitable for research applications.
  • Chemical identifiers: CAS 260367-12-2; molecular weight 341.36; formula C19H19NO5.