Learn More
Medchemexpress LLC Fmoc-NH-PEG1-CH2COOH | 260367-12-2 | MFCD06796876 | 99.9% | 341.36 g·mol⁻¹ | C19H19NO5 | 25 G
SDP
Catalog No. 5000203153
Encompass_Preferred
Fmoc-NH-PEG1-CH2COOH is an Fmoc-protected PEG1 linker bearing a terminal carboxylic acid used as a cleavable linker in antibody-drug conjugate (ADC) synthesis and as a PEG-based PROTAC linker in conjugation chemistry. It has CAS number 260367-12-2, molecular formula C19H19NO5, and a molecular weight of 341.36 g·mol⁻¹.
  • Cleavable ADC linker for antibody-drug conjugation.
  • Suitable as a PEG-based PROTAC linker.
  • Fmoc-protected amine enables standard peptide and solid-phase chemistries.
  • High reported purity suitable for synthetic applications.
  • Available in multiple pack sizes including 25 G for preparative work.
Catalog No. 50-002-03153 Supplier Medchemexpress LLC Supplier No. HYW05586125G
Add to Cart
Add to Cart
Fmoc-NH-PEG1-CH2COOH is an Fmoc-protected PEG1 linker bearing a terminal carboxylic acid used as a cleavable linker in antibody-drug conjugate (ADC) synthesis and as a PEG-based PROTAC linker in conjugation chemistry. It has CAS number 260367-12-2, molecular formula C19H19NO5, and a molecular weight of 341.36 g·mol⁻¹.
  • Cleavable ADC linker for antibody-drug conjugation.
  • Suitable as a PEG-based PROTAC linker.
  • Fmoc-protected amine enables standard peptide and solid-phase chemistries.
  • High reported purity suitable for synthetic applications.
  • Available in multiple pack sizes including 25 G for preparative work.