Learn More
Medchemexpress LLC HY-W055861 500mg Medchemexpress, Fmoc-NH-PEG1-CH2COOH CAS:260367-12-2 Purity:>98%
Supplier: Medchemexpress LLC HYW055861500MG
Medchemexpress, HY-W055861 500mg Fmoc-NH-PEG1-CH2COOH CAS:260367-12-2 Fmoc-NH-PEG1-CH2COOH is a cleavable ADC linker used in the synthesis of antibody-drug conjugates (ADCs). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.