Learn More
Medchemexpress LLC Fmoc-NH-PEG11-CH2CH2COOH | 3076038-46-2 | 98.5% | 50 MG
SDP
Catalog No. 5000340348
Encompass_Preferred
Fmoc-NH-PEG11-CH2CH2COOH is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one for an E3 ubiquitin ligase and the other for the target protein. They exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Peg-based protac linker
  • Used in the synthesis of protacs
  • Exploits intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-40348 Supplier Medchemexpress LLC Supplier No. HY13087050MG
Fmoc-NH-PEG11-CH2CH2COOH is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one for an E3 ubiquitin ligase and the other for the target protein. They exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Peg-based protac linker
  • Used in the synthesis of protacs
  • Exploits intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins