Learn More
eMolecules​ 2629308-66-1 | Fmoc-NH-PEG11-CH2COOH | Broadpharm | MFCD30723257 | 781.893 | C39H59NO15 | 95.000 | OC(=O)COCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCNC(=O)OCC1c2ccccc2-c2ccccc12 | 1g | 340377163
Catalog No. NC1692937 Shop All eMolecules Inc. Products
Encompass_Preferred
Fmoc-NH-PEG11-CH2COOH | Broadpharm | 2629308-66-1 | MFCD30723257 | 781.893 | C39H59NO15 | 95.000 | OC(=O)COCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCNC(=O)OCC1c2ccccc2-c2ccccc12 | 1g | 340377163
Catalog No. NC1692937 Supplier eMolecules​ Supplier No. 340377163
Fmoc-NH-PEG11-CH2COOH | Broadpharm | 2629308-66-1 | MFCD30723257 | 781.893 | C39H59NO15 | 95.000 | OC(=O)COCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCNC(=O)OCC1c2ccccc2-c2ccccc12 | 1g | 340377163