Learn More
Medchemexpress LLC Fmoc-NH-PEG2-CH2CH2COOH | 872679-70-4 | MFCD08064306 | 99.8% | 399.44 g/mol | C22H25NO6 | 10 G
SDP
Catalog No. 5000204271
Encompass_Preferred
Fmoc-NH-PEG2-CH2CH2COOH is an Fmoc-protected, monodisperse PEG2 linker with a terminal carboxylic acid. Supplied as a solid white powder, it is used as a cleavable linker for antibody-drug conjugates and as a PEG-based linker in PROTAC and conjugation chemistry.
  • Fmoc-protected amine for orthogonal deprotection.
  • Terminal carboxylic acid for conjugation or coupling.
  • Monodisperse PEG2 spacer to provide short flexible linkage.
  • High supplied purity (~99.8%) suitable for research use.
  • Solid form; store cold to preserve stability.
Catalog No. 50-002-04271 Supplier Medchemexpress LLC Supplier No. HYW04023810G
Fmoc-NH-PEG2-CH2CH2COOH is an Fmoc-protected, monodisperse PEG2 linker with a terminal carboxylic acid. Supplied as a solid white powder, it is used as a cleavable linker for antibody-drug conjugates and as a PEG-based linker in PROTAC and conjugation chemistry.
  • Fmoc-protected amine for orthogonal deprotection.
  • Terminal carboxylic acid for conjugation or coupling.
  • Monodisperse PEG2 spacer to provide short flexible linkage.
  • High supplied purity (~99.8%) suitable for research use.
  • Solid form; store cold to preserve stability.