Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical Fmoc-NH-PEG20-CH2CH2COOH | 1 g | CAS 1952360-93-8 | MDL MFCD21363316

Catalog No. 502343267
Encompass

Fmoc-NH-PEG20-CH2CH2COOH is a Amino Acid reagent (Subcategory: PEG AA) sold by WuXi TIDES. Offered in 1 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 1952360-93-8
- MDL: MFCD21363316
- InChIKey: YIKBHNBKAGGRHQ-UHFFFAOYSA-N
- Molecular Weight: 1192.397
- Molecular Formula: C58H97NO24
- Purity: ≥95%
- Container Type: 15 mL HDPE
- Pack Size: 1 g
- Net Weight: 1 g
- Gross Weight: 7.8 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: 1-(9H-fluoren-9-yl)-3-oxo-2,7,10,13,16,19,22,25,28,31,34,37,40,43,46,49,52,55,58,61,64-henicosaoxa-4-azaheptahexacontan-67-oic acid
- SMILES: O=C(CCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCNC(OCC1C2=C(C=CC=C2)C3=C1C=CC=C3)=O)O

Catalog No. 50-234-3267 Supplier STA Pharmaceutical Supplier No. STAAA119341GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-NH-PEG20-CH2CH2COOH is a Amino Acid reagent (Subcategory: PEG AA) sold by WuXi TIDES. Offered in 1 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 1952360-93-8
- MDL: MFCD21363316
- InChIKey: YIKBHNBKAGGRHQ-UHFFFAOYSA-N
- Molecular Weight: 1192.397
- Molecular Formula: C58H97NO24
- Purity: ≥95%
- Container Type: 15 mL HDPE
- Pack Size: 1 g
- Net Weight: 1 g
- Gross Weight: 7.8 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: 1-(9H-fluoren-9-yl)-3-oxo-2,7,10,13,16,19,22,25,28,31,34,37,40,43,46,49,52,55,58,61,64-henicosaoxa-4-azaheptahexacontan-67-oic acid
- SMILES: O=C(CCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCNC(OCC1C2=C(C=CC=C2)C3=C1C=CC=C3)=O)O

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.