Learn More
Medchemexpress LLC Fmoc-NH-PEG3-CH2CH2COOH | 867062-95-1 | 99.8% | 443.49 g/mol | C24H29NO7 | 10 G
SDP
Catalog No. 5000203181
Encompass_Preferred
Fmoc-NH-PEG3-CH2CH2COOH is an Fmoc-protected, PEG3-based linker used as a cleavable ADC linker and as a PEG-based PROTAC linker in synthetic and bioconjugation applications.
  • Fmoc-protected amino-PEG3 linker suitable for conjugation.
  • Used in ADC and PROTAC synthesis applications.
  • Molecular formula C24H29NO7 and molecular weight 443.49 g/mol.
  • High purity (99.8%) appropriate for research use.
  • Available in multiple pack sizes including 10 G.
Catalog No. 50-002-03181 Supplier Medchemexpress LLC Supplier No. HYW04023110G
Fmoc-NH-PEG3-CH2CH2COOH is an Fmoc-protected, PEG3-based linker used as a cleavable ADC linker and as a PEG-based PROTAC linker in synthetic and bioconjugation applications.
  • Fmoc-protected amino-PEG3 linker suitable for conjugation.
  • Used in ADC and PROTAC synthesis applications.
  • Molecular formula C24H29NO7 and molecular weight 443.49 g/mol.
  • High purity (99.8%) appropriate for research use.
  • Available in multiple pack sizes including 10 G.