Learn More
Medchemexpress LLC Fmoc-NH-PEG3-CH2CH2COOH | 867062-95-1 | 99.8% | 443.49 g/mol | C24H29NO7 | 5 G
SDP
Catalog No. 5000203102
Encompass_Preferred
Fmoc-NH-PEG3-CH2CH2COOH is an Fmoc-protected PEG3-based cleavable linker used for conjugation chemistry in research applications such as antibody-drug conjugates and PROTAC synthesis. It provides a short PEG spacer with an Fmoc-protected amino terminus and a terminal carboxylic acid for coupling reactions.
  • High purity suitable for research use.
  • PEG3 spacer provides solubility and flexibility.
  • Fmoc protection enables standard amine deprotection workflows.
  • Terminal carboxylic acid allows amide bond formation.
  • Available in multiple pack sizes for synthesis scale-up.
Catalog No. 50-002-03102 Supplier Medchemexpress LLC Supplier No. HYW0402315G
Fmoc-NH-PEG3-CH2CH2COOH is an Fmoc-protected PEG3-based cleavable linker used for conjugation chemistry in research applications such as antibody-drug conjugates and PROTAC synthesis. It provides a short PEG spacer with an Fmoc-protected amino terminus and a terminal carboxylic acid for coupling reactions.
  • High purity suitable for research use.
  • PEG3 spacer provides solubility and flexibility.
  • Fmoc protection enables standard amine deprotection workflows.
  • Terminal carboxylic acid allows amide bond formation.
  • Available in multiple pack sizes for synthesis scale-up.