Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fmoc-NH-PEG3-CH2CH2COOH | 867062-95-1 | MFCD06656471 | 99.8% | 443.49 g/mol | C24H29NO7 | 100 MG
SDP

Catalog No. 5000204177
Encompass_Preferred

Fmoc-NH-PEG3-CH2CH2COOH is a PEG-based, Fmoc-protected heterobifunctional linker designed for use in antibody-drug conjugate (ADC) and proteolysis-targeting chimera (PROTAC) construction. It features an Fmoc-protected amine and a terminal carboxylic acid that enable standard peptide-coupling and conjugation chemistries.

  • Fmoc-protected amine enabling orthogonal deprotection
  • Terminal carboxylic acid for amide coupling
  • Molecular formula C24H29NO7 and molecular weight 443.49 g/mol
  • Cas number 867062-95-1
  • Purity 99.8%
  • Available in small-scale packages such as 100 MG
  • Storage: pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months)
  • Suitable for ADC and PROTAC linker synthesis and bioconjugation

Catalog No. 50-002-04177 Supplier Medchemexpress LLC Supplier No. HYW040231100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-NH-PEG3-CH2CH2COOH is a PEG-based, Fmoc-protected heterobifunctional linker designed for use in antibody-drug conjugate (ADC) and proteolysis-targeting chimera (PROTAC) construction. It features an Fmoc-protected amine and a terminal carboxylic acid that enable standard peptide-coupling and conjugation chemistries.

  • Fmoc-protected amine enabling orthogonal deprotection
  • Terminal carboxylic acid for amide coupling
  • Molecular formula C24H29NO7 and molecular weight 443.49 g/mol
  • Cas number 867062-95-1
  • Purity 99.8%
  • Available in small-scale packages such as 100 MG
  • Storage: pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months)
  • Suitable for ADC and PROTAC linker synthesis and bioconjugation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.