Learn More
Medchemexpress LLC Fmoc-NH-PEG4-CH2COOH | 437655-95-3 | 99.1% | 473.52 | C25H31NO8 | 250 MG
SDP
Catalog No. 5000201984
Encompass_Preferred
Fmoc-NH-PEG4-CH2COOH is an Fmoc-protected PEG4 linker with a terminal carboxylic acid. It serves as a cleavable spacer in antibody-drug conjugate and PROTAC assembly, and as a pegylated spacer for peptide conjugation to increase solubility and provide a site for functionalization.
  • Fmoc-protected amine with terminal carboxylic acid
  • Peg4 spacer increases solubility and flexibility
  • Suitable for antibody-drug conjugate (ADC), proteolysis-targeting chimera (PROTAC), and peptide conjugation chemistries
  • Available in small-scale quantities for synthesis and method development
  • Store at low temperature in a sealed container to minimize moisture exposure
Catalog No. 50-002-01984 Supplier Medchemexpress LLC Supplier No. HY130175250MG
Fmoc-NH-PEG4-CH2COOH is an Fmoc-protected PEG4 linker with a terminal carboxylic acid. It serves as a cleavable spacer in antibody-drug conjugate and PROTAC assembly, and as a pegylated spacer for peptide conjugation to increase solubility and provide a site for functionalization.
  • Fmoc-protected amine with terminal carboxylic acid
  • Peg4 spacer increases solubility and flexibility
  • Suitable for antibody-drug conjugate (ADC), proteolysis-targeting chimera (PROTAC), and peptide conjugation chemistries
  • Available in small-scale quantities for synthesis and method development
  • Store at low temperature in a sealed container to minimize moisture exposure