Learn More
Medchemexpress LLC Fmoc-NH-PEG5-C2-NH2 | 2093277-71-3 | 98.1% | 502.60 g/mol | C27H38N2O7 | 5 MG
SDP
Catalog No. 5000223466
Encompass_Preferred
Fmoc-NH-PEG5-C2-NH2 is an Fmoc-protected PEG-based linker that provides a five-unit PEG spacer and a terminal primary amine for use in the synthesis of PROTACs and other bifunctional molecules. It is intended for medicinal chemistry applications where a flexible, hydrophilic linker and orthogonal protection are required.
  • Provides a five-unit polyethylene glycol spacer for flexibility and solubility.
  • Contains an Fmoc-protected amine for orthogonal deprotection strategies.
  • Includes a terminal primary amine for straightforward coupling to ligands or linkers.
  • Suitable for solution- or solid-phase synthesis workflows.
  • High purity suitable for research-grade synthesis.
Catalog No. 50-002-23466 Supplier Medchemexpress LLC Supplier No. HY1346825MG
Fmoc-NH-PEG5-C2-NH2 is an Fmoc-protected PEG-based linker that provides a five-unit PEG spacer and a terminal primary amine for use in the synthesis of PROTACs and other bifunctional molecules. It is intended for medicinal chemistry applications where a flexible, hydrophilic linker and orthogonal protection are required.
  • Provides a five-unit polyethylene glycol spacer for flexibility and solubility.
  • Contains an Fmoc-protected amine for orthogonal deprotection strategies.
  • Includes a terminal primary amine for straightforward coupling to ligands or linkers.
  • Suitable for solution- or solid-phase synthesis workflows.
  • High purity suitable for research-grade synthesis.