Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fmoc-NH-PEG5-CH2COOH | 635287-26-2 | MFCD26142982 | >97.0% | 517.57 g/mol | C27H35NO9 | 50 MG
SDP

Catalog No. 5000203694
Encompass_Preferred

Fmoc-NH-PEG5-CH2COOH is an Fmoc-protected PEG5 linker bearing a terminal carboxylic acid, supplied for research use and commonly used as a cleavable linker in antibody-drug conjugate (ADC) synthesis and as a PEG-based PROTAC linker.

  • Fmoc-protected amine with a terminal carboxyl functional group.
  • Peg5 spacer providing flexible, hydrophilic linkage.
  • Purity greater than or equal to 97.0%.
  • Molecular weight 517.57 g/mol.
  • CAS number 635287-26-2 for unambiguous identification.
  • Available in small-scale quantities; confirm specific pack size with supplier.

Catalog No. 50-002-03694 Supplier Medchemexpress LLC Supplier No. HY13306250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-NH-PEG5-CH2COOH is an Fmoc-protected PEG5 linker bearing a terminal carboxylic acid, supplied for research use and commonly used as a cleavable linker in antibody-drug conjugate (ADC) synthesis and as a PEG-based PROTAC linker.

  • Fmoc-protected amine with a terminal carboxyl functional group.
  • Peg5 spacer providing flexible, hydrophilic linkage.
  • Purity greater than or equal to 97.0%.
  • Molecular weight 517.57 g/mol.
  • CAS number 635287-26-2 for unambiguous identification.
  • Available in small-scale quantities; confirm specific pack size with supplier.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.