Learn More
Medchemexpress LLC Fmoc-NH-PEG6-CH2COOH | 437655-96-4 | 99.4% | 5 G
SDP
Catalog No. 5000339233
Encompass_Preferred
Fmoc-NH-PEG6-CH2COOH is a cleavable ADC linker used in the synthesis of antibody-drug conjugates (ADCs) and also a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. This compound leverages the intracellular ubiquitin-proteasome system for selective degradation of target proteins.
  • Cleavable ADC linker
  • PEG-based PROTAC linker
  • Utilized in antibody-drug conjugate (ADC) synthesis
  • Used in PROTAC synthesis
  • Leverages intracellular ubiquitin-proteasome system
Catalog No. 50-003-39233 Supplier Medchemexpress LLC Supplier No. HY1303645G
Fmoc-NH-PEG6-CH2COOH is a cleavable ADC linker used in the synthesis of antibody-drug conjugates (ADCs) and also a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. This compound leverages the intracellular ubiquitin-proteasome system for selective degradation of target proteins.
  • Cleavable ADC linker
  • PEG-based PROTAC linker
  • Utilized in antibody-drug conjugate (ADC) synthesis
  • Used in PROTAC synthesis
  • Leverages intracellular ubiquitin-proteasome system