Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fmoc-NH-PEG6-CH2COOH | 437655-96-4 | 99.4% | 561.62 | C29H39NO10 | 25 MG
SDP

Catalog No. 5000203491
Encompass_Preferred

Fmoc-NH-PEG6-CH2COOH is an Fmoc-protected PEG6 linker with a terminal carboxylic acid used as a hydrophilic spacer for conjugation chemistry. It is commonly used in antibody-drug conjugate (ADC) and PROTAC synthesis to provide a soluble linker and a protected amine for further derivatization.

  • Cleavable PEG-based linker suitable for ADC and PROTAC synthesis.
  • Provides an Fmoc-protected amine and a terminal carboxylic acid for conjugation.
  • Molecular weight 561.62 and formula C29H39NO10.
  • Purity 99.37% as reported by the supplier.
  • Soluble in DMSO at 100 mg/mL; may require sonication.
  • Storage: pure form -20 °C (3 years) or 4 °C (2 years); in solvent -80 °C (6 months) or -20 °C (1 month).
  • Available in small lab pack sizes, including 25 mg.

Catalog No. 50-002-03491 Supplier Medchemexpress LLC Supplier No. HY13036425MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Fmoc-NH-PEG6-CH2COOH is an Fmoc-protected PEG6 linker with a terminal carboxylic acid used as a hydrophilic spacer for conjugation chemistry. It is commonly used in antibody-drug conjugate (ADC) and PROTAC synthesis to provide a soluble linker and a protected amine for further derivatization.

  • Cleavable PEG-based linker suitable for ADC and PROTAC synthesis.
  • Provides an Fmoc-protected amine and a terminal carboxylic acid for conjugation.
  • Molecular weight 561.62 and formula C29H39NO10.
  • Purity 99.37% as reported by the supplier.
  • Soluble in DMSO at 100 mg/mL; may require sonication.
  • Storage: pure form -20 °C (3 years) or 4 °C (2 years); in solvent -80 °C (6 months) or -20 °C (1 month).
  • Available in small lab pack sizes, including 25 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.